siRNA Details
| Gene Symbol: 4930415L06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132812.1 | - | 2 | 363 | ||
| BC150985.1 | - | 1 | 314 | 9087898 | ||
| BC150987.1 | - | 1 | 314 | 9087900 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930415L06Rik| Gene Symbol: 4930415L06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132812.1 | - | 2 | 363 | ||
| BC150985.1 | - | 1 | 314 | 9087898 | ||
| BC150987.1 | - | 1 | 314 | 9087900 | ||