siRNA Details
Gene Symbol: Gm10825 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028580.1 | 2 | 2674 | |||
GenBank mRNA | AK148310.1 | - | 2 | 2607 | ||
AK163997.1 | - | 2 | 2606 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm10825 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028580.1 | 2 | 2674 | |||
GenBank mRNA | AK148310.1 | - | 2 | 2607 | ||
AK163997.1 | - | 2 | 2606 |