siRNA Details
Gene Symbol: Sec16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027641.1 | 1, 2 | 64 | |||
GenBank mRNA | BC059194.1 | - | 1, 2 | 50 | 4222915 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sec16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027641.1 | 1, 2 | 64 | |||
GenBank mRNA | BC059194.1 | - | 1, 2 | 50 | 4222915 |