siRNA Details
| Gene Symbol: 3830408C21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015471.1 | 1 | 290 | |||
| GenBank mRNA | AK014423.1 | - | 1 | 290 | ||
| AK138173.1 | - | 1 | 308 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3830408C21Rik| Gene Symbol: 3830408C21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015471.1 | 1 | 290 | |||
| GenBank mRNA | AK014423.1 | - | 1 | 290 | ||
| AK138173.1 | - | 1 | 308 | |||