siRNA Details
| Gene Symbol: Gm8615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028061.1 | 1 | 725 | |||
| GenBank mRNA | CB237974.1 | - | 1 | 121 | ||
| Gene Symbol: Gnpda1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF088903.1 | - | 7 | 900 | ||
| AF160355.1 | - | 7 | 888 | |||
| BC014800.1 | - | 7 | 873 | 4220980 | ||

Gm8615
Genomic Map