siRNA Details
| Gene Symbol: 5530601H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015467.1 | 5 | 3607 | |||
| GenBank mRNA | AK164650.1 | - | 5 | 3608 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5530601H04Rik| Gene Symbol: 5530601H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015467.1 | 5 | 3607 | |||
| GenBank mRNA | AK164650.1 | - | 5 | 3608 | ||