siRNA Details
| Gene Symbol: 5530601H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015467.1 | 5 | 1769 | |||
| GenBank mRNA | AK084653.1 | - | 6 | 1844 | ||
| AK084980.1 | - | 6 | 1823 | |||
| AK085098.1 | - | 6 | 1815 | |||
| AK164650.1 | - | 5 | 1770 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5530601H04Rik| Gene Symbol: 5530601H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015467.1 | 5 | 1769 | |||
| GenBank mRNA | AK084653.1 | - | 6 | 1844 | ||
| AK084980.1 | - | 6 | 1823 | |||
| AK085098.1 | - | 6 | 1815 | |||
| AK164650.1 | - | 5 | 1770 | |||