siRNA Details
| Gene Symbol: 1700084E18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028299.1 | 1 | 90 | |||
| GenBank mRNA | AK006990.1 | - | 1 | 91 | ||
| AK034708.1 | - | 684 | ||||
| BC062268.1 | - | 1 | 66 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700084E18Rik| Gene Symbol: 1700084E18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028299.1 | 1 | 90 | |||
| GenBank mRNA | AK006990.1 | - | 1 | 91 | ||
| AK034708.1 | - | 684 | ||||
| BC062268.1 | - | 1 | 66 | |||