siRNA Details
| Gene Symbol: Tmem80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024509.1 | 3 | 663 | |||
| GenBank mRNA | AK148367.1 | - | 3 | 634 | ||
| BC096570.1 | - | 3 | 617 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem80| Gene Symbol: Tmem80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024509.1 | 3 | 663 | |||
| GenBank mRNA | AK148367.1 | - | 3 | 634 | ||
| BC096570.1 | - | 3 | 617 | |||