siRNA Details
| Gene Symbol: A230072E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015600.2 | 3 | 2234 | |||
| GenBank mRNA | AK081752.1 | - | 3 | 2234 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230072E10Rik| Gene Symbol: A230072E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015600.2 | 3 | 2234 | |||
| GenBank mRNA | AK081752.1 | - | 3 | 2234 | ||