siRNA Details
| Gene Symbol: A130040M12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037717.1 | - | 1583 | |||
| AK149382.1 | - | 1 | 3188 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A130040M12Rik| Gene Symbol: A130040M12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037717.1 | - | 1583 | |||
| AK149382.1 | - | 1 | 3188 | |||