siRNA Details
| Gene Symbol: Vmn2r-ps159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028141.1 | 4 | 1556 | |||
| NR_038052.1 | 4 | 1403 | ||||
| GenBank mRNA | AF011418.1 | - | 4 | 1403 | ||
| AF011419.1 | - | 4 | 1556 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r-ps159| Gene Symbol: Vmn2r-ps159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028141.1 | 4 | 1556 | |||
| NR_038052.1 | 4 | 1403 | ||||
| GenBank mRNA | AF011418.1 | - | 4 | 1403 | ||
| AF011419.1 | - | 4 | 1556 | |||