siRNA Details
Gene Symbol: Vmn2r-ps159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028141.1 | 3, 4 | 1369 | |||
GenBank mRNA | AF011419.1 | - | 3, 4 | 1369 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn2r-ps159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028141.1 | 3, 4 | 1369 | |||
GenBank mRNA | AF011419.1 | - | 3, 4 | 1369 |