siRNA Details
| Gene Symbol: 1700086O06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015475.1 | 4 | 854 | |||
| NR_027903.1 | 2 | 560 | ||||
| NR_027904.1 | 2 | 463 | ||||
| GenBank mRNA | AK039796.1 | - | 2 | 464 | ||
| AK050707.1 | - | 4 | 829 | |||
| AK078369.1 | - | 2 | 560 | |||

1700086O06Rik
Genomic Map