siRNA Details
| Gene Symbol: 1700086O06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015475.1 | 4 | 1254 | |||
| NR_027903.1 | 2 | 960 | ||||
| NR_027904.1 | 2 | 863 | ||||
| GenBank mRNA | AK039796.1 | - | 2 | 864 | ||
| AK078369.1 | - | 2 | 960 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700086O06Rik| Gene Symbol: 1700086O06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015475.1 | 4 | 1254 | |||
| NR_027903.1 | 2 | 960 | ||||
| NR_027904.1 | 2 | 863 | ||||
| GenBank mRNA | AK039796.1 | - | 2 | 864 | ||
| AK078369.1 | - | 2 | 960 | |||