siRNA Details
| Gene Symbol: Rps4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003634.2 | 1 | 890 | |||
| GenBank mRNA | AK004068.1 | - | 1 | 891 | ||
| AK012382.1 | - | 1 | 890 | |||
| AK132160.1 | - | 1 | 891 | |||
| BC047994.1 | - | 1 | 862 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB204246.1 | - | 1 | 864 | Chr. 6 - 148354687 - 148355578 [+] on Build GRCm38 | |

Rps4l
Genomic Map