siRNA Details
| Gene Symbol: Snhg10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003145.3 | 2 | 378 | |||
| GenBank mRNA | AK006372.1 | - | 2 | 379 | ||
| BX524976.1 | - | 2 | 339 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snhg10| Gene Symbol: Snhg10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003145.3 | 2 | 378 | |||
| GenBank mRNA | AK006372.1 | - | 2 | 379 | ||
| BX524976.1 | - | 2 | 339 | |||