siRNA Details
Gene Symbol: Snhg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003145.3 | 2 | 356 | |||
GenBank mRNA | AK006372.1 | - | 2 | 357 | ||
BX524976.1 | - | 2 | 317 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Snhg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003145.3 | 2 | 356 | |||
GenBank mRNA | AK006372.1 | - | 2 | 357 | ||
BX524976.1 | - | 2 | 317 |