siRNA Details
| Gene Symbol: C330024D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015582.2 | 5 | 2362 | |||
| GenBank mRNA | AK049327.1 | - | 5 | 2363 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C330024D21Rik| Gene Symbol: C330024D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015582.2 | 5 | 2362 | |||
| GenBank mRNA | AK049327.1 | - | 5 | 2363 | ||