siRNA Details
| Gene Symbol: Il4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027491.1 | 1 | 14 | |||
| GenBank mRNA | AA199406.1 | - | 1 | 14 | ||
| BX529138.1 | - | 1 | 23 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il4| Gene Symbol: Il4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027491.1 | 1 | 14 | |||
| GenBank mRNA | AA199406.1 | - | 1 | 14 | ||
| BX529138.1 | - | 1 | 23 | |||