siRNA Details
| Gene Symbol: 4930594C11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024017.1 | 1 | 229 | |||
| GenBank mRNA | AK019839.1 | - | 1 | 229 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930594C11Rik| Gene Symbol: 4930594C11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024017.1 | 1 | 229 | |||
| GenBank mRNA | AK019839.1 | - | 1 | 229 | ||