siRNA Details
| Gene Symbol: 4930404I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028368.1 | 2 | 108 | |||
| GenBank mRNA | AK006191.1 | - | 2 | 99 | ||
| AK015081.1 | - | 2 | 110 | |||
| AK029496.1 | - | 1 | 1564 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930404I05Rik| Gene Symbol: 4930404I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028368.1 | 2 | 108 | |||
| GenBank mRNA | AK006191.1 | - | 2 | 99 | ||
| AK015081.1 | - | 2 | 110 | |||
| AK029496.1 | - | 1 | 1564 | |||