siRNA Details
| Gene Symbol: 4930404I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028368.1 | 3 | 783 | |||
| GenBank mRNA | AK006191.1 | - | 3 | 774 | ||
| AK015081.1 | - | 3 | 785 | |||
| AK029496.1 | - | 2 | 2239 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714611.1 | - | 3 | 781 | Chr. 16 - 91011251 - 91016441 [+] on Build GRCm38 | |

4930404I05Rik
Genomic Map