siRNA Details
| Gene Symbol: 4930404I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028368.1 | 3 | 361 | |||
| GenBank mRNA | AK006191.1 | - | 3 | 352 | ||
| AK015081.1 | - | 3 | 363 | |||
| AK029496.1 | - | 2 | 1817 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930404I05Rik| Gene Symbol: 4930404I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028368.1 | 3 | 361 | |||
| GenBank mRNA | AK006191.1 | - | 3 | 352 | ||
| AK015081.1 | - | 3 | 363 | |||
| AK029496.1 | - | 2 | 1817 | |||