siRNA Details
| Gene Symbol: Gm1943 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002928.2 | 1 | 1074 | |||
| GenBank mRNA | AK170003.1 | - | 1 | 1082 | ||
| BC024407.1 | - | 1 | 1075 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm1943| Gene Symbol: Gm1943 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002928.2 | 1 | 1074 | |||
| GenBank mRNA | AK170003.1 | - | 1 | 1082 | ||
| BC024407.1 | - | 1 | 1075 | |||