siRNA Details
Gene Symbol: Gm1943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002928.2 | 1 | 582 | |||
GenBank mRNA | AK170003.1 | - | 1 | 590 | ||
BC024407.1 | - | 1 | 583 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm1943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002928.2 | 1 | 582 | |||
GenBank mRNA | AK170003.1 | - | 1 | 590 | ||
BC024407.1 | - | 1 | 583 |