siRNA Details
| Gene Symbol: 8430429K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028317.1 | 3 | 596 | |||
| GenBank mRNA | AK086952.1 | - | 3 | 561 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_027820.1 | - | 561 | Chr. - - [-] on Build | ||

8430429K09Rik
Genomic Map