siRNA Details
| Gene Symbol: 8430429K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028317.1 | 1 | 288 | |||
| NR_045276.1 | 1 | 242 | ||||
| GenBank mRNA | AK086952.1 | - | 1 | 253 | ||
| BY069744.1 | - | 1 | 243 | |||
| BY331368.1 | - | 1 | 245 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8430429K09Rik| Gene Symbol: 8430429K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028317.1 | 1 | 288 | |||
| NR_045276.1 | 1 | 242 | ||||
| GenBank mRNA | AK086952.1 | - | 1 | 253 | ||
| BY069744.1 | - | 1 | 243 | |||
| BY331368.1 | - | 1 | 245 | |||