siRNA Details
Gene Symbol: 8430429K09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028317.1 | 2 | 514 | |||
GenBank mRNA | AK086952.1 | - | 2 | 479 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_027820.1 | - | 479 | Chr. - - [-] on Build |
