siRNA Details
| Gene Symbol: Atp10d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003966.1 | 16 | 3406 | |||
| GenBank mRNA | AK036646.1 | - | 15 | 3054 | ||
| AK133052.1 | - | 5 | 843 | |||
| BC141328.1 | - | 16 | 3243 | 9055940 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp10d| Gene Symbol: Atp10d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003966.1 | 16 | 3406 | |||
| GenBank mRNA | AK036646.1 | - | 15 | 3054 | ||
| AK133052.1 | - | 5 | 843 | |||
| BC141328.1 | - | 16 | 3243 | 9055940 | ||