siRNA Details
| Gene Symbol: Bmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033241.1 | 16 | 2437 | |||
| GenBank mRNA | AK147534.1 | - | 14 | 3353 | ||
| BC003925.1 | - | 6 | 766 | |||
| BC035204.1 | - | 4 | 508 | |||
| BC053923.1 | - | 16 | 2327 | |||
| L35280.1 | - | 2 | 81 | |||

Bmp1
Genomic Map