siRNA Details
| Gene Symbol: Bmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033241.1 | 16 | 2893 | |||
| GenBank mRNA | AK147534.1 | - | 14 | 3809 | ||
| BC003925.1 | - | 6 | 1222 | |||
| BC053923.1 | - | 16 | 2784 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmp1| Gene Symbol: Bmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033241.1 | 16 | 2893 | |||
| GenBank mRNA | AK147534.1 | - | 14 | 3809 | ||
| BC003925.1 | - | 6 | 1222 | |||
| BC053923.1 | - | 16 | 2784 | |||