siRNA Details
| Gene Symbol: Gm12216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033332.1 | 4 | 699 | |||
| GenBank mRNA | AK138811.1 | - | 4 | 700 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF466244.1 | - | 4 | 588 | Chr. 11 - 53785921 - 53859144 [-] on Build GRCm38 | |

Gm12216
Genomic Map