siRNA Details
| Gene Symbol: Gm12216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033332.1 | 4 | 907 | |||
| GenBank mRNA | AK138811.1 | - | 4 | 908 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12216| Gene Symbol: Gm12216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033332.1 | 4 | 907 | |||
| GenBank mRNA | AK138811.1 | - | 4 | 908 | ||