siRNA Details
| Gene Symbol: C87499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027385.1 | 1, 2 | 71 | |||
| GenBank mRNA | AK163216.1 | - | 1, 2 | 73 | ||
| CJ083078.1 | - | 1, 2 | 71 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C87499| Gene Symbol: C87499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027385.1 | 1, 2 | 71 | |||
| GenBank mRNA | AK163216.1 | - | 1, 2 | 73 | ||
| CJ083078.1 | - | 1, 2 | 71 | |||