siRNA Details
| Gene Symbol: 1700030C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015521.1 | 2 | 731 | |||
| GenBank mRNA | AK006529.1 | - | 2 | 519 | ||
| BC100365.1 | - | 2 | 731 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700030C10Rik| Gene Symbol: 1700030C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015521.1 | 2 | 731 | |||
| GenBank mRNA | AK006529.1 | - | 2 | 519 | ||
| BC100365.1 | - | 2 | 731 | |||