siRNA Details
Gene Symbol: Mir124a-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029814.1 | 1 | 52 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NR_031867.
Gene Ontology Categories:
