siRNA Details
| Gene Symbol: Tug1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002321.2 | 3 | 1123 | |||
| NR_002322.2 | 2 | 1519 | ||||
| NR_110490.1 | 2 | 1519 | ||||
| GenBank mRNA | AK033381.1 | - | 2 | 1491 | ||
| AK048213.1 | - | 2 | 1511 | |||
| AK083955.1 | - | 2 | 1503 | |||
| AK088174.1 | - | 3 | 1124 | |||
| BC016239.1 | - | 2 | 868 | |||
| BC048828.1 | - | 2 | 1395 | |||
| DQ517434.1 | - | 2 | 1520 | |||
| DQ517435.1 | - | 3 | 1119 | |||

Tug1
Genomic Map