siRNA Details
| Gene Symbol: Tug1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002321.2 | 3 | 1014 | |||
| NR_002322.2 | 2 | 1410 | ||||
| NR_110490.1 | 2 | 1410 | ||||
| GenBank mRNA | AK033381.1 | - | 2 | 1382 | ||
| AK048213.1 | - | 2 | 1402 | |||
| AK083955.1 | - | 2 | 1394 | |||
| AK088174.1 | - | 3 | 1015 | |||
| BC016239.1 | - | 2 | 759 | |||
| BC048828.1 | - | 2 | 1286 | |||
| DQ517434.1 | - | 2 | 1411 | |||
| DQ517435.1 | - | 3 | 1010 | |||

Tug1
Genomic Map