siRNA Details
| Gene Symbol: Tug1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002321.2 | 3 | 1259 | |||
| NR_002322.2 | 2 | 1655 | ||||
| NR_110490.1 | 2 | 1655 | ||||
| GenBank mRNA | AK033381.1 | - | 2 | 1627 | ||
| AK048213.1 | - | 2 | 1647 | |||
| AK088174.1 | - | 3 | 1260 | |||
| BC016239.1 | - | 2 | 1004 | |||
| BC048828.1 | - | 2 | 1531 | |||
| DQ517434.1 | - | 2 | 1656 | |||
| DQ517435.1 | - | 3 | 1255 | |||

Tug1
Genomic Map