siRNA Details
| Gene Symbol: 1700001G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033199.1 | 1 | 143 | |||
| GenBank mRNA | AK005592.1 | - | 1 | 143 | ||
| AK008236.1 | - | 1 | 126 | |||
| AK008325.1 | - | 1 | 126 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB575681.1 | - | 1 | 109 | Chr. 1 - 33669883 - 33670722 [+] on Build GRCm38 | |

1700001G17Rik
Genomic Map