siRNA Details
| Gene Symbol: Snora69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002900.1 | 1 | 40 | |||
| GenBank mRNA | AF357420.1 | - | 1 | 40 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snora69| Gene Symbol: Snora69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002900.1 | 1 | 40 | |||
| GenBank mRNA | AF357420.1 | - | 1 | 40 | ||