siRNA Details
| Gene Symbol: Gm11149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029465.1 | 5, 6 | 596 | |||
| GenBank mRNA | AK015955.1 | - | 5, 6 | 597 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715520.1 | - | 5, 6 | 597 | Chr. 9 - 49518288 - 49567661 [+] on Build GRCm38 | |

Gm11149
Genomic Map