siRNA Details
| Gene Symbol: Gm11149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029465.1 | 6 | 1195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11149| Gene Symbol: Gm11149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029465.1 | 6 | 1195 | |||