siRNA Details
| Gene Symbol: Mir713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030493.1 | 1 | 21 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mir713| Gene Symbol: Mir713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030493.1 | 1 | 21 | |||