siRNA Details
| Gene Symbol: Mir713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030493.1 | 1 | 94 | |||
| Gene Symbol: 6720490N10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020176.1 | - | 1 | 31 | ||

Mir713
Genomic Map