siRNA Details
| Gene Symbol: C230004F18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030706.1 | 5 | 281 | |||
| GenBank mRNA | AK082088.1 | - | 262 | |||
| AK163488.1 | - | 5 | 274 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_177916.1 | - | 262 | Chr. - - [-] on Build | ||

C230004F18Rik
Genomic Map