siRNA Details
| Gene Symbol: C230004F18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030706.1 | 8 | 675 | |||
| GenBank mRNA | AK163488.1 | - | 8 | 668 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C230004F18Rik| Gene Symbol: C230004F18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030706.1 | 8 | 675 | |||
| GenBank mRNA | AK163488.1 | - | 8 | 668 | ||