siRNA Details
| Gene Symbol: ChkbCpt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004843.2 | 10, 11 | 1785 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ChkbCpt1b| Gene Symbol: ChkbCpt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004843.2 | 10, 11 | 1785 | |||