siRNA Details
| Gene Symbol: A930007I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015567.2 | 4 | 473 | |||
| GenBank mRNA | AK137192.1 | - | 4 | 475 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB627860.1 | - | 4 | 475 | Chr. 19 - 29505303 - 29521989 [-] on Build GRCm38 | |

A930007I19Rik
Genomic Map